Skip to Content
Merck
All Photos(1)

Documents

SAAR1123420

Ammonium molybdate tetrahydrate

univAR

Synonym(s):

Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)6Mo7O24 · 4H2O
CAS Number:
Molecular Weight:
1235.86
MDL number:
UNSPSC Code:
12352302

product line

univAR

packaging

poly bottle of 12 × 500 g (SAAR1123420EM)

density

2.498 g/mL at 25 °C (lit.)

SMILES string

N.N.N.N.N.N.O.O.O.O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O

InChI

1S/7Mo.6H3N.10H2O.18O/h;;;;;;;6*1H3;10*1H2;;;;;;;;;;;;;;;;;;/q;;;;3*+2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-6

InChI key

FIXLYHHVMHXSCP-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

Analyzed reagent
For laboratory use only.

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service