20479
GlasSeal Capillary Column Connector, Borosilicate Glass
pkg of 12 ea
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 12 ea
technique(s)
gas chromatography (GC): suitable
Related Categories
General description
Column connectors are useful for attaching a guard column/retention gap to an analytical column, or for repairing a broken column.
Quickly connect two pieces of fused silica tubing of the same or different diameters without the need for tools. The leak-free connection is useful to connect a guard column or transfer line, repair a broken column, or connect two columns (same or different phases). Silanized for an inert inside surface. For use with 0.20 - 0.53 mm I.D. fused silica tubing.
Other Notes
We offer a variety of chromatography accessories including analytical syringes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)