Skip to Content
Merck

T124

Thapsigargin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C34H50O12
CAS Number:
Molecular Weight:
650.75
UNSPSC Code:
12352204
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

storage temp.

−20°C

SMILES string

[H][C@@]12C([C@H](OC([C@]3(O)C)=O)[C@]3(O)[C@@H](OC(CCC)=O)C[C@]2(C)OC(C)=O)=C(C)[C@H](OC(/C(C)=C(C)/[H])=O)[C@H]1OC(CCCCCCC)=O

InChI

1S/C34H50O12/c1-9-12-13-14-15-17-24(37)43-28-26-25(20(5)27(28)44-30(38)19(4)11-3)29-34(41,33(8,40)31(39)45-29)22(42-23(36)16-10-2)18-32(26,7)46-21(6)35/h11,22,26-29,40-41H,9-10,12-18H2,1-8H3/b19-11-/t22-,26+,27-,28-,29-,32-,33+,34+/m0/s1

InChI key

IXFPJGBNCFXKPI-FSIHEZPISA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potent, cell-permeable, IP3-independent intracellular calcium releaser.
Thapsigargin is a potent, cell-permeable, IP3-independent intracellular calcium release. It blocks the transient increase in intracellular Ca2+ induced by angiostatin and endostatin. It also induces apoptosis by disrupting intracellular free Ca2+ levels and is incorporated into chemotherapeutic prodrug formulations.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ridwan Babatunde Ibrahim et al.
Cellular and molecular life sciences : CMLS, 77(7), 1421-1434 (2019-11-16)
Transthyretin amyloidosis (ATTR) is a progressive life-threatening disease characterized by the deposition of transthyretin (TTR) amyloid fibrils. Several pathogenic variants have been shown to destabilize TTR tetramers, leading to aggregation of misfolded TTR fibrils. However, factors that underlie the differential
Wencheng He et al.
Frontiers in microbiology, 8, 2129-2129 (2017-11-23)
Classical swine fever (CSF) is an OIE-listed, highly contagious animal disease caused by classical swine fever virus (CSFV). The endoplasmic reticulum (ER) is an organelle in which the replication of many RNA viruses takes place. During viral infection, a series
Moran Dvela-Levitt et al.
Cell, 178(3), 521-535 (2019-07-28)
Intracellular accumulation of misfolded proteins causes toxic proteinopathies, diseases without targeted therapies. Mucin 1 kidney disease (MKD) results from a frameshift mutation in the MUC1 gene (MUC1-fs). Here, we show that MKD is a toxic proteinopathy. Intracellular MUC1-fs accumulation activated
Macarena Pavez et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(26), 5095-5114 (2019-04-27)
The spatial and temporal regulation of calcium signaling in neuronal growth cones is essential for axon guidance. In growth cones, the endoplasmic reticulum (ER) is a significant source of calcium signals. However, it is not clear whether the ER is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service