Skip to Content
Merck

SAB4200367

Anti-FLVCR1 antibody, Mouse monoclonal

clone FL-58, purified from hybridoma cell culture

Synonym(s):

Anti-AXPC1, Anti-FLVCR, Anti-Ffline leukemia virus subgroup C cellular receptor 1, Anti-MFSD7B, Anti-PCA, Anti-PCARP

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

NACRES:
NA.41
UNSPSC Code:
51111800
Conjugate:
unconjugated
Clone:
FL-58, monoclonal
Application:
IF, WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

FL-58, monoclonal

form

buffered aqueous solution

mol wt

antigen ~72 kDa

species reactivity

human, rat, monkey, mouse

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2.5-5 μg/mL using A549 cells, western blot: 2.0-4.0 μg/mL using extracts of Raji cells

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FLVCR1(28982)

General description

Feline leukemia virus subgroup C receptor 1 (FLVCR1) is a member of cell surface receptors that bind feline leukemia virus, subgroup C.
Monoclonal Anti-FLVCR1 (mouse IgG2a isotype) is derived from the hybridoma FL-58 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a peptide corresponding to the C-terminus of human FLVCR1 conjugated to KLH.

Immunogen

peptide corresponding to the C-terminus of human FLVCR1, conjugated to KLH. The corresponding sequence is identical in monkey and bovine FLVCR1.

Application

Anti-FLVCR1 antibody may be used for immunoblotting at a working concentration of 2.0-4.0 μg/ml using Raji cell extracts. The antibody is suitable for immunofluorescence at a recommended concentration of 2.5-5.0 μg/ml using A549 cells.
Monoclonal Anti-FLVCR1 antibody produced in mouse has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Monoclonal Anti-FLVCR1 recognizes human, monkey, mouse and rat FLVCR1 (~72 kDa).
Human feline leukemia virus subgroup C receptor 1 (FLVCR1) is required to transport cytoplasmic heme and maintain intracellular heme concentration, that protects the developing erythroid cells from heme toxicity. Disruption of FLVCR1 is implicated in fatal congenital anemia called Diamond-Blackfan anemia, degeneration of neurons in retinitis pigmentosa and posterior column ataxia via dysregulation of heme homeostasis). Knockdown of FLVCR is associated with defective erythropoiesis and disturbed systemic iron homeostasis in mice. Feline leukemia viruses (FeLVs) are pathogenic retroviruses of cats that disrupt the function of host receptor and causes fatal anemia.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutations in FLVCR1 cause posterior column ataxia and retinitis pigmentosa
Rajadhyaksha AM, et al.
American Journal of Human Genetics, 87(5), 643-654 (2010)
Control of intracellular heme levels: heme transporters and heme oxygenases
Khan AA and Quigley JG
Biochimica et Biophysica Acta - Molecular Cell Research, 1813(5), 668-682 (2011)
Identification of a human heme exporter that is essential for erythropoiesis
Quigley JG, et al.
Cell, 118(6), 757-766 (2004)
Jennifer K Brown et al.
Journal of virology, 80(4), 1742-1751 (2006-01-28)
Infection of cells by the highly anemogenic feline leukemia virus subgroup C (FeLV-C) is mediated by the heme exporter FLVCR1, a cell surface protein containing 12 potential transmembrane segments with six presumptive extracellular loops (ECLs). To identify FLVCR1 residues critical

Related Content

Global Trade Item Number

SKUGTIN
SAB4200367-200UL04061838036100

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service