Skip to Content
Merck
All Photos(1)

Documents

PZ0333

Sigma-Aldrich

PF-06412154 hydrochloride

≥98% (HPLC)

Synonym(s):

N-[4-[(1R)-1-[(3-Methyl-2-quinolinyl)amino]butyl]benzoyl]-β-alanine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27N3O3 · HCl
CAS Number:
Molecular Weight:
441.95
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

[H]Cl.OC(CCNC(C1=CC=C([C@H](NC2=C(C)C=C(C=CC=C3)C3=N2)CCC)C=C1)=O)=O

InChI

1S/C24H27N3O3.ClH/c1-3-6-20(26-23-16(2)15-19-7-4-5-8-21(19)27-23)17-9-11-18(12-10-17)24(30)25-14-13-22(28)29;/h4-5,7-12,15,20H,3,6,13-14H2,1-2H3,(H,25,30)(H,26,27)(H,28,29);1H/t20-;/m1./s1

InChI key

BTOHFLWOXYLUQK-VEIFNGETSA-N

Biochem/physiol Actions

PF-06412154 hydrochloride is a glucagon receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service