Skip to Content
Merck

P2149

Physalaemin

≥97% (HPLC)

Synonym(s):

pGlu-Ala-Asp-Pro-Asn-Lys-Phe-Tyr-Gly-Leu-Met-NH2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C58H84N14O16S
CAS Number:
Molecular Weight:
1265.44
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(CC(C)C)NC(=O)CNC(=O)C(Cc1ccc(O)cc1)NC(=O)C(Cc2ccccc2)NC(=O)C(CCCCN)NC(=O)C(CC(N)=O)NC(=O)C3CCCN3C(=O)C(CC(O)=O)NC(=O)C(C)NC(=O)C4CCC(=O)N4)C(N)=O

Gene Information

human ... INS(3630)

Biochem/physiol Actions

Potent hypotensive; induces salivation, intestinal contraction and vasodilation

Other Notes

Tachykinin originally isolated from the skin of the amphibian, Physalaemus fuscumaculatus; with structural homology to substance P




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Takahiko Shiina et al.
European journal of pharmacology, 628(1-3), 202-206 (2009-12-05)
We examined the effects of physalaemin, an agonist of tachykinin receptors, on mechanical responses in the rat esophagus to clarify possible regulatory roles of tachykinins in esophageal motility. Exogenous application of physalaemin caused tonic contractions in rat esophageal segments when
Daniel A Nelson et al.
Frontiers in bioscience : a journal and virtual library, 9, 2166-2176 (2004-09-09)
Mammalian tachykinins are traditionally viewed as neuropeptides. This review describes the mammalian tachykinins and evidence for expression of these peptides by non-neuronal cells. Tachykinin expression is defined as evidence for gene transcription, peptide production, or peptide secretion. Since the functions
Christy Rani R Grace et al.
Biopolymers, 96(3), 252-259 (2010-07-16)
Physalaemin (PHY), a non-mammalian tachykinin, binds selectively to neurokinin-1 (NK1) receptor with high affinity. Both the aqueous and lipid-induced conformations of PHY have been studied using two-dimensional nuclear magnetic resonance techniques. These data show that in water PHY prefers to