Skip to Content
Merck
All Photos(2)

Documents

MAK227

Sigma-Aldrich

Chloramphenicol Extraction Kit

greener alternative

sufficient for 40 extractions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

cosmetics
food and beverages
sample preservation

greener alternative category

storage temp.

room temp

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product reduces solvent waste over traditional methods. Find details here.

Application

The Chloramphenicol Extraction Kit is intended for the extraction of chloramphenicol from food and biological samples. It can be used for any biological food sample - shrimp, meat, liver, crab, fish, etc.

Click on the following link for a video demonstration of how the kit works

Features and Benefits

The Chloramphenicol Extraction kit shortcuts the extraction process by eliminating the need for centrifugation and manual pipetting. After that, the extract can be used to detect chloramphenicol by ELISA and spectrophotometric analysis.

Suitability

This kit is suitable for the extraction of chloramphenicol from a variety of samples such as biological tissue and food.

Principle

Chloramphenicol, or chlornitromycin, is a broad-spectrum antibiotic that is used to treat a wide variety of bacterial infections. However, due to potential serious side effects its use has been banned in many countries. Minimum acceptable limits have been set for chloramphenicol residues in food samples. This chloramphenicol extraction kit enables the separation of chloramphenicol from tissue and tissue water components. The extracted chloramphenicol can then be detected and quantified by enzyme-linked immunosorbent assay (ELISA) followed by spectrophotometric analysis. In the standard extraction, chloramphenicol is typically extracted using ethyl acetate. In this procedure, ethyl acetate containing the chloramphenicol is separated from tissue by centrifugation for 10 minutes followed by manual pipetting. An aliquot is then dried and reconstituted into 1 mL each of iso-octane:chloroform (2:3) solution and sample dilution buffer, of which 50 μL of sample dilution buffer is used for analysis by ELISA and spectrophotometry.

comparable product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

3 - Flammable liquids

Flash Point(F)

10.4 °F

Flash Point(C)

-12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service