Skip to Content
Merck

L8280

Latex beads, carboxylate-modified, biotin-labeled, yellow-green

buffered aqueous suspension, 0.05 μm mean particle size

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

buffered aqueous suspension

composition

Solids, 1.0%

mean particle size

0.05 μm

Physical form

Suspension in 10 mM PBS, pH 7.1, 2 mM NaN3




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library