Skip to Content
Merck

L4628

Linoleic anhydride

~97%, oil

Synonym(s):

(cis,cis)-9,12-Octadecadienoic anhydride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C36H62O3
CAS Number:
Molecular Weight:
542.88
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~97%

form

oil

color

clear colorless

refractive index

n20/D 1.476 (lit.)

mp

−1 °C (lit.)

density

0.901 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/CCCCCCCC(=O)OC(=O)CCCCCCC\C=C/C\C=C/CCCCC



pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library