Skip to Content
Merck

L0380

Lumazine

crystalline

Synonym(s):

2,4(1H,3H)-Pteridinedione, 2,4-Dihydroxypteridine, 2,4-Pteridinediol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H4N4O2
CAS Number:
Molecular Weight:
164.12
EC Number:
207-652-7
UNSPSC Code:
12352204
MDL number:
Beilstein/REAXYS Number:
157503
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

crystalline

color

yellow

SMILES string

Oc1nc(O)c2nccnc2n1

InChI

1S/C6H4N4O2/c11-5-3-4(8-2-1-7-3)9-6(12)10-5/h1-2H,(H2,8,9,10,11,12)

InChI key

UYEUUXMDVNYCAM-UHFFFAOYSA-N



Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library