Skip to Content

F1508

N-Fmoc-O-t-Butyl-L-threonine

Synonym(s):

Fmoc-Thr(tBu)-OH, Fmoc-O-tert-butyl-L-threonine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H27NO5
CAS Number:
Molecular Weight:
397.46
EC Number:
276-261-1
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
4581133
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

−20°C

SMILES string

C[C@@H](OC(C)(C)C)[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C23H27NO5/c1-14(29-23(2,3)4)20(21(25)26)24-22(27)28-13-19-17-11-7-5-9-15(17)16-10-6-8-12-18(16)19/h5-12,14,19-20H,13H2,1-4H3,(H,24,27)(H,25,26)/t14-,20+/m1/s1

InChI key

LZOLWEQBVPVDPR-VLIAUNLRSA-N



Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library