Skip to Content
Merck

D2391

N,N′-Diallyl L-tartardiamide

Synonym(s):

DATD, N,N′-Diallyltartramide

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C10H16N2O4
CAS Number:
Molecular Weight:
228.25
UNSPSC Code:
12352000
EC Number:
261-277-3
Beilstein/REAXYS Number:
1712934
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

storage temp.

2-8°C

SMILES string

O[C@H]([C@@H](O)C(=O)NCC=C)C(=O)NCC=C

Application

DATD is a reversible cross-linker for polyacrylamide gels that produces a gel which is soluble in dilute (2%) periodic acid at room temperature for 1-2 hr. Gel solubilization is useful when recovering protein from polyacrylamide gels.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A method for the quantitative determination of protein incorporated in solubilized polyacrylamide gels.
J Tas et al.
Analytical biochemistry, 100(2), 264-270 (1979-12-01)
Quantitative measurement of protein mass and radioactivity in N,N'-diallyltartardiamide crosslinked polyacrylamide slab gels.
R B Young et al.
Analytical biochemistry, 108(1), 202-206 (1980-10-01)
Polyacrylamide gels with modified cross-linkages.
P B O'Connell et al.
Analytical biochemistry, 76(l), 63-73 (1976-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service