Documents
CLS6798410D
Corning® stirrer with digital display
AC/DC input 230 V AC (CEE7-7 plug), plate L × W 5 in. × 7 in.
Synonym(s):
magnetic stirrer, magnetic stirrers, stirrers
About This Item
UNSPSC Code:
41103803
NACRES:
NB.73
packaging
case of 1
manufacturer/tradename
Corning 6798-410D
availability
not available in USA, Canada and EU
parameter
60-1150 rpm speed
plate L × W
5 in. × 7 in.
General description
  • Digital LED stir speed display indicates when set speed has been reached
  • Microprocessor maintains consistent and repeatable stir speed settings
  • Extremely durable and heat resistant Pyroceram glass-ceramic top
  • Small footprint and low profile
  • Meets UL and cUL standards
For use in Asia, Africa, and Australia.
Other Notes
individual units are not routinely calibrated, rpm speeds are approximate only
Legal Information
Corning is a registered trademark of Corning, Inc.