Skip to Content
Merck
All Photos(1)

Documents

CLS3658

Corning® 384 well microplate

black polypropylene, round bottom, non-sterile, lid: no, pack of 25

Synonym(s):

384 multiwell plate, 384 well fluorescent assay plate, 384 well microplate, 384 well microtiter plate, 384 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

black bottom
black polypropylene
round bottom
square wells

sterility

non-sterile

feature

lid: no
skirt
microplate for fluorescent assays
plate format: 384 well standard

packaging

case of 100
pack of 25

manufacturer/tradename

Corning 3658

size

384 wells

well volume

95 μL

well working volume

15-70 μL

color

black

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 384 well microplates support various application requirements.
  • Non-treated clear polystyrene surface is suitable primarily for the immobilization of large molecules, such as antibodies.
  • The round bottom is usually the best choice for washing, retrieving and sample auto-centering.
  • Ideal for fluorescent assays requiring solvent resistance (e.g., DMSO, ethanol, methanol)
  • Conforms to standard microplate footprint and dimensions
  • Total well volume 95 μL; working volume 15-70 μL.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service