Skip to Content
Merck

C5265

4-Formylbenzonitrile

≥95% (GC)

Synonym(s):

4-Cyanobenzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NCC6H4CHO
CAS Number:
Molecular Weight:
131.13
EC Number:
203-267-3
UNSPSC Code:
12352100
MDL number:
Beilstein/REAXYS Number:
606473
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (GC)

bp

133 °C/12 mmHg (lit.)

mp

100-102 °C (lit.)

storage temp.

2-8°C

SMILES string

O=Cc1ccc(cc1)C#N

InChI

1S/C8H5NO/c9-5-7-1-3-8(6-10)4-2-7/h1-4,6H

InChI key

WZWIQYMTQZCSKI-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library