Skip to Content

C3876

Z-Ala-Phe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H22N2O5
CAS Number:
Molecular Weight:
370.40
UNSPSC Code:
12352200
EC Number:
220-454-5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


SMILES string

C[C@H](NC(=O)OCc1ccccc1)C(=O)N[C@@H](Cc2ccccc2)C(O)=O



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library