Skip to Content
Merck

B003

Chloroethylclonidine dihydrochloride

solid

Synonym(s):

2-[2,6-Dichloro(N-β-chloroethyl-N-methyl)-4-aminomethyl]phenylimino-2-imidazolidine dihydrochloride, CEC dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C13H17Cl3N4 · 2HCl
CAS Number:
Molecular Weight:
408.58
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Chloroethylclonidine dihydrochloride, solid

InChI

1S/C13H17Cl3N4.2ClH/c1-20(5-2-14)8-9-6-10(15)12(11(16)7-9)19-13-17-3-4-18-13;;/h6-7H,2-5,8H2,1H3,(H2,17,18,19);2*1H

SMILES string

Cl.Cl.CN(CCCl)Cc1cc(Cl)c(\N=C2/NCCN2)c(Cl)c1

InChI key

CZSAXNDACBPMAB-UHFFFAOYSA-N

form

solid

color

off-white

solubility

0.1 M NaOH: 1 mg/mL (Stable for 24 hours at 4 °C.)
H2O: 12 mg/mL (Stable for 24 hours at 4 °C.)
methanol: 17 mg/mL (Stable for 24 hours at 4 °C.)
aqueous base: unstable

Gene Information

human ... ADRA1B(147)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Irreversible α1B-adrenoceptor alkylating agent.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Leclerc et al.
British journal of pharmacology, 71(1), 5-9 (1980-01-01)
1 alpha-Adrenoceptor stimulant activity was determined for noradrenaline (NA), clonidine and a series of para-substituted derivatives of clonidine on rat aortic strips, a rat brain synaptosome preparation, and anaesthetized pithed rats. The effects on the blood pressure of intraventricular (i.c.v.)
L Xiao et al.
European journal of pharmacology, 347(2-3), 319-327 (1998-07-08)
We quantified and compared the rates at which chloroethylclonidine (CEC) inactivated cloned rat alpha1A, alpha1B-, and alpha1D-adrenoceptors. Membranes from cells transfected with one of the three cloned alpha1-adrenoceptors were incubated for various intervals with 100 microM chloroethylclonidine at 10 degrees
Yoav Litvin et al.
The European journal of neuroscience, 40(2), 2344-2351 (2014-05-28)
Research suggests a causal link between estrogens and mood. Here, we began by examining the effects of estradiol (E2 ) on rat innate and conditioned defensive behaviors in response to cat odor. Second, we utilized whole-cell patch clamp electrophysiological techniques
M Takanashi et al.
Naunyn-Schmiedeberg's archives of pharmacology, 343(6), 669-673 (1991-06-01)
The influence of the alpha 1b-adrenoceptor-selective antagonist chlorethylclonidine on the alpha 1-adrenergic positive inotropic effect and the phosphoinositide hydrolysis induced by phenylephrine was investigated in the rabbit ventricular myocardium. Pretreatment of membrane fractions derived from the rabbit ventricular muscle with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service