Skip to Content
Merck

Skip To

AV49429

Anti-TMCO1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HP10122, Anti-PCIA3, Anti-PNAS-136, Anti-RP11-466F5.7, Anti-TMCC4, Anti-Transmembrane and coiled-coil domains 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Please contact Customer Service for Availability
2.300,00 PLN

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
3

2.300,00 PLN


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

21 kDa

species reactivity

mouse, bovine, horse, rat, dog, rabbit, human, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMCO1(54499)

Immunogen

Synthetic peptide directed towards the C terminal region of human TMCO1

Application

Anti-TMCO1 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

Transmembrane and coiled-coil domains 1 (TMCO1) is a membrane protein of DUF841 superfamily. While the function of this protein is not clear, homozygous frameshift mutations in this gene causes craniofacial dysmorphism, skeletal anomalies, and mental retardation.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: CSFIFLYILCTMSIRQNIQKILGLAPSRAATKQAGGFLGPPPPSGKFS

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA054768HPA053894HPA021643
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

technique(s)

western blot: suitable

technique(s)

immunofluorescence: 0.25-2 μg/mL

technique(s)

immunohistochemistry: 1:20- 1:50

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:20-1:50

Gene Information

human ... TMCO1(54499)

Gene Information

human ... TMCO1(54499)

Gene Information

human ... TMCC1(23023)

Gene Information

human ... CCDC44(51204)


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters