Skip to Content
Merck

A7097

Austricine

≥98% (TLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H18O4 · H2O
Molecular Weight:
280.32
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (TLC)

storage temp.

2-8°C

SMILES string

C[C@H]1[C@@H]2[C@@H](O)CC(C)=C3C([C@H]2OC1=O)C(C)=CC3=O

Biochem/physiol Actions

A hypolipidemic, antiatherosclerotic, anti-inflammatory agent, with angioprotective and hepatoprotective activity; antiulcerous.

Other Notes

The molecular formula of Austricine, by definition, includes one water of hydration. The name should not be listed as "hydrate."

Disclaimer

Protect from light.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library