Skip to Content
Merck

66030

Adenosine 5′-triphosphate disodium salt

Synonym(s):

Adenosine 5′-triphosphate (ATP) disodium salt hydrate, ATP disodium hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H14N5O13P3Na2 · xH2O
CAS Number:
Molecular Weight:
551.14 (anhydrous basis)
UNSPSC Code:
41106305
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

packaging

vial of 30 mg

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O

InChI

1S/C10H16N5O13P3.2Na.H2O/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;;1H2/q;2*+1;/p-2/t4-,6-,7-,10-;;;/m1.../s1

InChI key

NTBQNWBHIXNPRU-MSQVLRTGSA-L

Biochem/physiol Actions

P2 purinergic agonist; increases activity of Ca2+-activated K+ channels; substrate for ATP-dependent enzyme systems

Packaging

Preweighed vials

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service