Skip to Content
Merck

5107

ICAM2 human

recombinant, expressed in E. coli, 0.5 mg protein/mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.75
UNSPSC Code:
12352202
Biological source:
human
Recombinant:
expressed in E. coli
Assay:
≥90% (SDS-PAGE)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

recombinant

expressed in E. coli

description

0.1 mg recombinant human ICAM2 in 20 mM Tris-HCl buffer, containing NaCl, KCl, EDTA, L-arginine, DTT and glycerol.

sterility

Filtered sterilized solution

assay

≥90% (SDS-PAGE)

form

liquid

packaging

pkg of 100 μg

concentration

0.5 mg protein/mL

accession no.

NP_000864

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... ICAM2(3384)

Application

Coating a plate well (6 well plate) with this recombinant ICAM2 protein in neuronal cell specific medium at 1-10 μg / well allows for 1) human lymphocyte cell / receptor interaction studies in vitro and 2) use as a culture matrix protein for anti-tumor immuno-response studies in vitro.

Use these recommendations as guidelines to determine the optimal coating conditions for your culture system.
1. Thaw ICAM2 and dilute to desired concentration using serum-free medium or PBS. The final solution should be sufficiently dilute so that the volume added covers the surface evenly.
Note: Use 1 ml PBS per well in a 6-well plate.
2. Add 1 - 10 μg protein to each well and incubate at 2-10 °C overnight.
3. After incubation, aspirate remaining material.
4. Plates are ready for use. They may also be stored at 2-8 °C damp or air dried if sterility is maintained.

Preparation Note

The full-length extracellular domain of human ICAM2 gene (25-223 aa) was constructed with 29 N-terminal T7/HIS-tag and expressed in E. coli as inclusion bodies. The final product was refolded using a unique “temperature shift inclusion body refolding” technology and chromatographically purified.

Other Notes

MASMTGGQQMGRGHHHHHHGNLYFQGGEFELKVFEVHVRPKKLAVEPKGSLEVNCSTTCNQPEVGGLETSLDKILLDEQAQWKHYLVSNISHDTVLQCHFTCSGKQESMNSNVSVYQPPRQVILTLQPTLVAVGKSFTIECRVPTVEPLDSLTLFLFRGNETLHYETFGKAAPAPQEATATFNSTADREDGHRNFSCLAVLDLMSRGGNIFHKHSAPKMLEIYEPVSDSQ


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library