Skip to Content
Merck
All Photos(1)

Documents

44687

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl β-D-ribofuranoside

≥98.0% (HPLC), powder

Synonym(s):

X β-D-ribofuranoside, X-riboside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13BrClNO5
CAS Number:
Molecular Weight:
378.60
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

5-Bromo-4-chloro-3-indolyl β-D-ribofuranoside, ≥98.0% (HPLC)

Assay

≥98.0% (HPLC)

form

powder

optical activity

[α]/D -102.0±3.0°, c = 1 in methanol

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3ccc(Br)c(Cl)c23)[C@H](O)[C@@H]1O

InChI

1S/C13H13BrClNO5/c14-5-1-2-6-9(10(5)15)7(3-16-6)20-13-12(19)11(18)8(4-17)21-13/h1-3,8,11-13,16-19H,4H2/t8-,11-,12-,13-/m1/s1

InChI key

RYLVNYDICPAOQR-HUXSOILUSA-N

Application

Evaluation of novel beta-ribosidase substrates for the differentiation of Gram-negative bacteria

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Supplied by Glycosynth Limited, U.S. Patent No. 7,323,488, European Patent Nos. 1436411 and 1438424, for use only in internal research conducted by the purchaser. No rights under the aforementioned patents are conveyed which permit the resale, transfer, or use for purposes other than research. Rights under the aforementioned patents, for purposes other than internal research, may be obtained by contacting Glycosynth Limited.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L A Butterworth et al.
Journal of applied microbiology, 96(1), 170-176 (2003-12-18)
To synthesize novel substrates for the detection of beta-ribosidase and assess their potential for the differentiation of Gram-negative bacteria. Two novel chromogenic substrates, 3',4'-dihydroxyflavone-4'-beta-D-ribofuranoside (DHF-riboside) and 5-bromo-4-chloro-3-indolyl-beta-D-ribofuranoside (X-riboside) were evaluated along with a known fluorogenic substrate, 4-methylumbelliferyl-beta-D-ribofuranoside (4MU-riboside). A total

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service