Skip to Content
Merck

41022

Atto 495 maleimide

BioReagent, suitable for fluorescence

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

form

powder

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 500 nm; λem 524 nm in 0.1 M phosphate pH 7.0

suitability

corresponds to standard for coupling to thiols, suitable for fluorescence

storage temp.

−20°C

SMILES string

[O-]Cl(=O)(=O)=O.CN(C)c1ccc2C=C3C=C\C(C=C3N(CCCC(=O)NCCN4C(=O)C=CC4=O)c2c1)=[N+](\C)C

InChI

1S/C27H31N5O3.ClHO4/c1-29(2)21-9-7-19-16-20-8-10-22(30(3)4)18-24(20)31(23(19)17-21)14-5-6-25(33)28-13-15-32-26(34)11-12-27(32)35;2-1(3,4)5/h7-12,16-18H,5-6,13-15H2,1-4H3;(H,2,3,4,5)

InChI key

IEQSJFQKROSLEM-UHFFFAOYSA-N

Application

Atto fluorescent labels are designed for high sensitivity applications, including single molecule detection. Atto labels have rigid structures that do not show any cis-trans-isomerization. Thus these labels display exceptional intensity with minimal spectral shift on conjugation. Atto 495 maleimide is suitable for fluorescence labeling of thiol/SH containing molecules.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library