Skip to Content
Merck
All Photos(1)

Key Documents

13767

Sigma-Aldrich

(S)-(−)-N-Benzyl-α-methylbenzylamine

puriss., ≥98.5% (sum of enantiomers, GC)

Synonym(s):

(S)-(−)-N-(1-Phenylethyl)benzylamine, (S)-(−)-N-Benzyl-α-phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NHCH2C6H5
CAS Number:
Molecular Weight:
211.30
Beilstein:
3650264
MDL number:
UNSPSC Code:
12352000

grade

puriss.

Assay

≥98.5% (sum of enantiomers, GC)

optical activity

[α]20/D −58±2°, c = 5% in ethanol

refractive index

n20/D 1.563 (lit.)

bp

171 °C/15 mmHg (lit.)

density

1.01 g/mL at 25 °C (lit.)

SMILES string

C[C@H](NCc1ccccc1)c2ccccc2

InChI

1S/C15H17N/c1-13(15-10-6-3-7-11-15)16-12-14-8-4-2-5-9-14/h2-11,13,16H,12H2,1H3/t13-/m0/s1

InChI key

ZYZHMSJNPCYUTB-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Silica gel orange, which is functionalized with xylenol orange, may be used as a selective solid phase extractor for the preconcentration and removal of mercury in water, which can be analyzed by spectrophotometric methods. It may also be used as a support in chelating resins prepared from amberlite XAD-2 and titron, which can find applications in preconcentration and determination of metal ions, via spectrophotometric methods.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of xylenol orange functionalized silica gel as a selective solid phase extractor and its application for preconcentration?separation of mercury from waters
Fan J, et al.
Journal of Hazardous Materials, 145(1), 323-330 (2007)
Metal ion enrichment with Amberlite XAD-2 functionalized with Tiron: analytical applications
Kumar M, et al.
Analyst, 125(6), 1221-1226 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service