Skip to Content
Merck
All Photos(1)

Key Documents

82814

Sigma-Aldrich

Pyridinium 3-nitrobenzenesulfonate

purum, ≥98.0% (T)

Synonym(s):

3-Nitrobenzenesulfonic acid pyridine salt, PNBS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N · C6H5NO5S
CAS Number:
Molecular Weight:
282.27
Beilstein:
5685497
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥98.0% (T)

mp

119-122 °C (lit.)

SMILES string

c1ccncc1.OS(=O)(=O)c2cccc(c2)[N+]([O-])=O

InChI

1S/C6H5NO5S.C5H5N/c8-7(9)5-2-1-3-6(4-5)13(10,11)12;1-2-4-6-5-3-1/h1-4H,(H,10,11,12);1-5H

InChI key

WZZFZXZRKPNZOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Mild acid catalyst used for the formation of cyclic orthoester, used e.g. as protecting groups in nucleosides

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Sekine et al.
Journal of the American Chemical Society, 105, 2044-2044 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service