Skip to Content
Merck

79535

5-Phospho-D-ribose 1-diphosphate pentasodium salt

≥80% (enzymatic)

Synonym(s):

α-D-Ribosyl diphosphate 5-phosphate pentasodium salt, 5-Phospho-α-D-ribosyl diphosphate, 5-Phosphoribosyl 1-pyrophosphate pentasodium salt, P-Rib-PP, PRPP pentasodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H8Na5O14P3
CAS Number:
Molecular Weight:
499.98
UNSPSC Code:
12352201
Beilstein/REAXYS Number:
6043637
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥80% (enzymatic)

shelf life

limited shelf life

impurities

≤18% water, ≤4% solvent

solubility

H2O: 50 mg/mL, clear, colorless to very faintly yellow

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@H](O[C@@H]1COP(O)([O-])=O)OP([O-])(=O)OP([O-])([O-])=O

InChI

1S/C5H13O14P3.4Na/c6-3-2(1-16-20(8,9)10)17-5(4(3)7)18-22(14,15)19-21(11,12)13;;;;/h2-7H,1H2,(H,14,15)(H2,8,9,10)(H2,11,12,13);;;;/q;4*+1/p-4/t2-,3-,4-,5?;;;;/m1..../s1

InChI key

QZAPPJHVNYCTAX-CKJQBBATSA-J





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library