Skip to Content
Merck
All Photos(2)

Documents

45989

Supelco

3-Nitroaniline

analytical standard

Synonym(s):

3-Aminonitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NH2
CAS Number:
Molecular Weight:
138.12
Beilstein:
636962
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

vapor pressure

1 mmHg ( 119 °C)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

111-114 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Nc1cccc(c1)[N+]([O-])=O

InChI

1S/C6H6N2O2/c7-5-2-1-3-6(4-5)8(9)10/h1-4H,7H2

InChI key

XJCVRTZCHMZPBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Nitroaniline belongs to the class of aromatic amines, found to be highly mutagenic and carcinogenic. These aromatic amines are widely used in the manufacturing of cosmetics, medicines, dyes and as intermediates in chemical syntheses.

Application

3-Nitroaniline may be used as an analytical reference standard for the quantification of the analyte in environmental samples using high-performance liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Heart,Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

384.8 °F - closed cup

Flash Point(C)

196.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 3-nitroaniline in water samples by directly suspended droplet three-phase liquid-phase microextraction using 18-crown-6 ether and high-performance liquid chromatography
Yazdi SA, et al.
Journal of Chromatography A, 1216(26), 5086-5091 (2009)
Analysis of aromatic amines in water samples by liquid-liquid-liquid microextraction with hollow fibers and high-performance liquid chromatography
Zhao L, et al.
Journal of Chromatography A, 963(1-2), 239-248 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service