Skip to Content
Merck

14201

Ammonium metavanadate

puriss., ≥98.5%

Synonym(s):

Ammonium vanadate(V)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NH4VO3
CAS Number:
Molecular Weight:
116.98
EC Number:
232-261-3
UNSPSC Code:
12352300
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

puriss.

assay

≥98.5%

density

2.32 g/cm3 at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg, phosphate (PO43-): ≤100 mg/kg

cation traces

Fe: ≤50 mg/kg, Pb: ≤50 mg/kg

SMILES string

N.O[V](=O)=O

InChI

1S/H3N.H2O.2O.V/h1H3;1H2;;;/q;;;;+1/p-1

InChI key

YBVKNHXQSRDWAA-UHFFFAOYSA-M



Still not finding the right product?

Explore all of our products under Ammonium metavanadate


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - STOT RE 1 Inhalation

target_organs

Respiratory Tract

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library