Skip to Content
Merck
All Photos(1)

Key Documents

NA14

Sigma-Aldrich

Anti-Topoisomerase II (Ab-1) Mouse mAb (SWT3D1)

liquid, clone SWT3D1, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody product type

primary antibodies

clone

SWT3D1, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

should not react with

rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with P3X63Ag8.653 mouse myeloma cells. Recognizes the ~170 kDa topoisomerase II protein.
Recognizes ~170 kDa topoisomerase II protein in HeLa cells and breast carcinoma tissue.

Immunogen

recombinant human topoisomerase IIα

Application

Immunoblotting (5 µg/ml, see application references)

Paraffin Sections (2.5 µg/ml, pressure cooker pre-treatment required)

Immunoprecipitation (not recommended)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM sodium phosphate buffer, 2% gelatin.

Analysis Note

Negative Control
P388D cells
Positive Control
HeLa cells or breast carcinoma tissue

Other Notes

Has a high affinity for topoisomerase IIα and a low to moderate affinity to topoisomerase IIβ. Antibody should be titrated for optimal results in individual systems.

van der Zee, A.G., et al. 1994. Ann. Oncol.5, 75.
Cummings, J. and Smyth, J.F. 1993. Ann. Oncol.4, 533.
Robinson, R.G., et al. 1993. Hybridoma12, 407.
Anderson, H.J. and Roberge, M. 1992. Cell Biol. Intl. Reports16, 717.
Gasser, S.M., et al. 1992. J. Antonie Van Leeuwenhoek62, 15.
Tan, K.B., et al. 1992. Can. Res.52, 231.
Osheroff, N., et al. 1991. Bioessays13, 269.
Chung, T.D.Y., et al. 1989. Proc. Natl. Acad. Sci. USA86, 9431.
Wang, J.C. 1985. Ann. Rev. Biochem.54, 665.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michele R Plewes et al.
Biology of reproduction, 101(5), 1001-1017 (2019-07-28)
Yes-associated protein 1 (YAP1) is a major component of the Hippo signaling pathway. Although the exact extracellular signals that control the Hippo pathway are currently unknown, increasing evidence supports a critical role for the Hippo pathway in embryonic development, regulation
Saumitri Bhattacharyya et al.
The Journal of biological chemistry, 284(22), 14966-14977 (2009-03-31)
The BLM helicase associates with the telomere structural proteins TRF1 and TRF2 in immortalized cells using the alternative lengthening of telomere (ALT) pathways. This work focuses on identifying protein partners of BLM in cells using ALT. Mass spectrometry and immunoprecipitation
Lasse Tengbjerg Hansen et al.
International journal of cancer, 105(4), 472-479 (2003-04-25)
Etoposide (VP16) is a potent inducer of DNA double-strand breaks (DSBs) and is efficiently used in small cell lung cancer (SCLC) therapy. However, acquired VP16 resistance remains an important barrier to effective treatment. To understand the underlying mechanisms for VP16
Mitchell R Harancher et al.
Journal of virology, 94(19) (2020-07-24)
Lysine-specific demethylase 1 (LSD1) targets cellular proteins, including histone H3, p53, E2F, and Dnmt1, and is involved in the regulation of gene expression, DNA replication, the cell cycle, and the DNA damage response. LSD1 catalyzes demethylation of histone H3K9 associated
Aurélia E Lewis et al.
Molecular & cellular proteomics : MCP, 10(2), M110-M110 (2010-11-05)
Considerable insight into phosphoinositide-regulated cytoplasmic functions has been gained by identifying phosphoinositide-effector proteins. Phosphoinositide-regulated nuclear functions however are fewer and less clear. To address this, we established a proteomic method based on neomycin extraction of intact nuclei to enrich for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service