Merck
Skip To
MASVFCASE
Portable case
For use with MAS-100 VF®
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 unit
Estimated to ship on28 July 2026fromKuehne + Nagel Sp. z o.o.
2.210,00 PLN
About This Item
NACRES:
JA.11
UNSPSC Code:
41103403
Feature:
portable
2.210,00 PLN
Estimated to ship on28 July 2026Details
Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
feature
portable
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with MAS-100 VF®
General description
The Portable case is an additional accessory for MAS-100 VF®, used for safe transport of the equipment. The carrying case provides space for one MAS-100 VF® air sampler including perforated lid and dust cover, USB memory stick, allen key, battery charger and documentation sheets such as Feller table and calibration certificates.
Application
The Portable case is used for safe carrying of MAS-100 VF® sampler that finds application in air sampling in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications.
Legal Information
MAS-100 VF is a registered trademark of MBV AG
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
1.441501.173341.73020
description
For use with MAS-100 VF®
description
for use with RCS® High Flow Touch
description
for use with MAS-100 Atmos®
description
-
feature
portable
feature
portable
feature
portable
feature
-
compatibility
for use with MAS-100 VF®
compatibility
for use with RCS® High Flow Touch
compatibility
for use with MAS-100 Atmos®
compatibility
for use with Spectroquant® Prove 100, for use with Spectroquant® Prove 300, for use with Spectroquant® Prove 600
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
application(s)
compressed gas monitoring
food and beverages
pharmaceutical
application(s)
-
Choose from one of the most recent versions:
Certificates of Analysis (COA)