Skip to Content
Merck
All Photos(1)

Documents

MAFCN0B50

Millipore

Multiscreen GFC Filter Plate, Opaque Non-sterile

Receptor/ligand binding assays, sample prep,TCA precipitation

Synonym(s):

GFC Filter Plate, Opaque GFC Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.22

material

Barex®
PVDF membrane (hydrophilic)
flat bottom wells
glass fiber membrane
titanium dioxide

Quality Level

sterility

non-sterile

product line

MultiScreen®

feature

hydrophilic
lid

manufacturer/tradename

MultiScreen®

parameter

50-250 μL sample volume (per well)

technique(s)

ligand binding assay: suitable
protein purification: suitable (TCA precipitation)
receptor binding assay: suitable

H

14.4 mm

L

128 mm

W

85.5 mm

filtration area

0.26 cm2

plate size

96 wells

well maximum volume

300 μL

working volume

50-250 μL

application(s)

sample preparation

binding type

high binding surface

shipped in

ambient

General description

Filter Code: FC
Glass Fiber Filter Membrane Filters:
Glass fiber filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated or particular laden samples. They are also widely used for clarification of aqueous solutions.
Glass fiber filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated or particulate-laden samples. They are also widely used for clarification of aqueous solutions.

Features & Benefits:
- Multiple configurations and applications
- Chemical compatibility
- Automation-compatible
- Superior vacuum filtration and filtrate collection

Applications:
- Receptor/Ligand Binding Assays, Sample Preparation, TCA Precipitation of Protein

Application

Research Category
General Filtration

Assays

Physical form

Color Code: Opaque

Other Notes

Automation Compatible: No

Legal Information

Barex is a registered trademark of INEOS Barex AG
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service