Skip to Content
Merck
All Photos(1)

Documents

8.55043

Sigma-Aldrich

IBX polystyrene

Novabiochem®

Synonym(s):

Polystyrene-Bound IBX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111023
NACRES:
NA.22

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: alcohol reactive

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

General description

This polymer-bound version of IBX cleanly converts primary and secondary alcohols to the corresponding aldehydes and ketones [1,2]. It is particularly useful for the synthesis of peptide aldehydes as it oxidises peptide alcohols efficiently and spent resin can be simply removed by filtration.

Literature references

[1] G. Sorg, et al. (2001) Angew. Chem. Int. Ed., 40, 4395.
[2] G. Sorg, et al. (2005) J. Pept. Sci.,11, 147.

Application

  • 2-Iodoxybenzoic Acid: An Oxidant for Functional Group Transformations: This study discusses the use of IBX for various organic transformations, including the oxidation of silyl enol ethers and the preparation of polystyrene supported periodinane reagent. (Nair, 2020).

Linkage

Replaces: 01-64-0445

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined by elemental analysis of iodine): 0.90 - 1.50 mmol/g
Oxidation of Cinnamyl alcohol to Cinnamaldehyde (2 eq. IBX PS): ≥ 90 %
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly (styrene - 1% DVB) 200 -400 mesh.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service