Skip to Content
Merck

676483

VEGFR Tyrosine Kinase Inhibitor IV - CAS 475108-18-0 - Calbiochem

The VEGFR Tyrosine Kinase Inhibitor IV, also referenced under CAS 475108-18-0, controls the biological activity of VEGFR Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

VEGFR Tyrosine Kinase Inhibitor IV - CAS 475108-18-0 - Calbiochem, Vascular Endothelial Growth Factor Receptor Tyrosine Kinase Inhibitor IV, N-(2-Chloro-4-((6,7-dimethoxy-4-quinolyl)oxy)phenyl)-Nʹ-(5-methyl-3-isoxazolyl)urea, PDGFR Tyrosine Kinase Inhibitor XII, VEGFR2 Kinase Inhibitor XXI, VEGFR1 Kinase Inhibitor, Vascular Endothelial Growth Factor Receptor Tyrosine Kinase Inhibitor IV, N-(2-Chloro-4-((6,7-dimethoxy-4-quinolyl)oxy)phenyl)-Nʹ-(5-methyl-3-isoxazolyl)urea, PDGFR Tyrosine Kinase Inhibitor XII, VEGFR2 Kinase Inhibitor XXI, VEGFR1 Kinase Inhibitor V

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H19ClN4O5 · xH2O
CAS Number:
Molecular Weight:
454.86 (anhydrous basis)
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

pink

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable quinoline-urea compound that potently inhibits the kinase activity of VEGFR (IC50 = 30, 6.5, and 15 nM for VEGFR-1,-2, and -3, respectively), EphB2, PDGFR-α, PDGFR-β, c-Kit, and Tie-2 (IC50 = 24, 40, 49, 78, and 78 nM, respectively). It exhibits little or no activity towards a panel of 15 other tyrosine kinases, including FGFR, IR, and IGFR, in cell-free kinase assays, while showing more than 7-fold higher potency in inhibiting ligand-induced phosphorylation of VEGFR-1/2/3 vs. c-Kit, PDGFR-β, Flt3, FGFR1, and c-Met in cell-based assays. Shown to be orally available and suppress tumor angiogenesis and growth in rats in vivo.
A cell-permeable quinoline-urea compound that potently inhibits the kinase activity of VEGFR (IC50 = 30, 6.5, and 15 nM for VEGFR-1,-2,and -3, respectively), EphB2, PDGFR-α, PDGFR-β, c-Kit, and Tie-2 (IC50 = 24, 40, 49, 78, and 78 nM, respectively). It exhibits little or no activity towards a panel of 15 other tyrosine kinases, including FGFR, IR, and IGFR, in cell-free kinase assays, while showing more than 7-fold higher potency in inhibiting ligand-induced phosphorylation of VEGFR-1/2/3 vs. c-Kit, PDGFR-β, Flt3, FGFR1, and c-Met in cell-based assays. Shown to be orally available and suppress tumor angiogenesis and growth in rats in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
VEGFR
Product does not compete with ATP.
Reversible: no
Target IC50: 30, 6.5, and 15 nM for VEGFR-1,-2, and -3, respectively; 24, 40, 49, 78, and 78 nM for EphB2, PDGFR-α, PDGFR-β, c-Kit, and Tie-2, respectively

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Nakamura, K., et al. 2006. Cancer Res.66, 9134.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

10-13 - German Storage Class 10 to 13



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library