Skip to Content
Merck

Skip To

5.89587

Chloroform

anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer

Synonym(s):

Methylidyne trichloride, Trichloromethane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Please contact Customer Service for Availability
PLN 373.00
1 L
Please contact Customer Service for Availability
PLN 619.00
2 L
Please contact Customer Service for Availability
PLN 1,270.00

About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
NACRES:
NA.22
UNSPSC Code:
12190000
Beilstein/REAXYS Number:
1731042
MDL number:
Assay:
≥99%
Grade:
anhydrous
Bp:
60.5-61.5 °C (lit.)
Vapor pressure:
160 mmHg ( 20 °C)

PLN 373.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

anhydrous

Quality Segment

vapor density

4.1 (vs air)

vapor pressure

160 mmHg ( 20 °C)

assay

≥99%

contains

0.5-1.0% ethanol as stabilizer

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
49618924216366927
Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

100

grade

anhydrous

grade

biotech. grade

grade

-

grade

HPLC grade

assay

≥99%

assay

≥99.8%

assay

99-99.4% (GC)

assay

≥99.8%

refractive index

n20/D 1.445 (lit.)

refractive index

n20/D 1.445 (lit.)

refractive index

n20/D 1.445 (lit.)

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

bp

60.5-61.5 °C (lit.)

bp

60-62 °C, 60.5-61.5 °C (lit.)

bp

60.5-61.5 °C (lit.)

vapor pressure

160 mmHg ( 20 °C)

vapor pressure

160 mmHg ( 20 °C)

vapor pressure

160 mmHg ( 20 °C)

vapor pressure

160 mmHg ( 20 °C)


Still not finding the right product?

Explore all of our products under Chloroform


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

wgk

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-66-3

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters