Skip to Content
Merck

370700

GW9662 - CAS 22978-25-2 - Calbiochem

Synonym(s):

2-Chloro-5-nitro-N-phenylbenzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (HPLC)

form

crystalline solid

solubility

DMSO: 20 mg/mL, methanol: soluble

General description

A cell-permeable, selective and irreversible PPARγ antagonist (IC50 = 3.3 nM, 32 nM, and 2 μM for PPARγ, PPARα, and PPARδ, respectively). Reported to covalently modify a cysteine residue in the binding site of PPAR. At a concentration of 10 μM, also acts as an agonist of human pregnane X receptor (PXR) and farnesoid X receptor (FXR). Does not activate liver X receptor-α (LXRα), retinoic acid receptor (RAR), retinoid X receptor-α (RXRα) and thyroid receptors α and β (TRα and TRβ).

Biochem/physiol Actions

Target IC50:3.3 nM, 32 nM, and 2 μM for PPARγ, PPARα, and PPARΔ

Preparation Note

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.


Storage Class

10-13 - German Storage Class 10 to 13



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library