Skip to Content
Merck
All Photos(1)

Key Documents

1.46505

Millipore

Inactivator Sol. w. LTHTh

Sign Into View Organizational & Contract Pricing


About This Item

sterility

sterile; autoclaved

Quality Level

form

liquid

feature

ready-to-use

pH

7 ( in H2O)

application(s)

bioburden testing
cosmetics
food and beverages
pharmaceutical

suitability

nonselective for

Packaging

90 ml in 275 ml wide mouth bottle (6 bottles per box)

Storage and Stability

Store at +2°C to +25°C.

Other Notes

Customized product – please contact your Sales representative for further information on purchase conditions.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service