Skip to Content
Merck
All Photos(1)

Documents

1.46321

Millipore

Bile Esculin Azide Agar

Ready-to-use 90mm plate for confirmation of intestinal Enterococci by Membrane filtration method.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

Agency

ISO 7899

Quality Level

sterility

sterile

form

plate

feature

ready-to-use

packaging

pkg of (90 mm plate (20 plates per box))

technique(s)

microbiological culture: suitable

pH

7.1 ( in H2O)

application(s)

bioburden testing
environmental
food and beverages
microbiology
water monitoring

suitability

Enterococcus spp.

General description

Bile Esculin Azide Agar are stackable specially designed plates that helps increase the safety during handling, transport and incubation. The agar plates are of standard 90mm diameter. Bile esculin azide agar is a selective media for isolation and presumptive identification of intestinal Enterococci by Membrane filtration method. Casein, peptone, yeast extract serves as sources of carbon, nitrogen, amino acids, vitamins and essential growth nutrients. Bile and sodium azide, inhibits most of the other accompanying bacteria. Esculin in the medium is hydrolyzed to esculetin and dextrose. Esculetin reacts with ammonium ferric citrate to form a dark brown or black complex, visualized as a zone of black precipitate around the colonies.

Application

Bile Esculin agar is used for the confirmation of pathogenic Enterococcus sp in water samples as stated in the ISO 7899.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service