Skip to Content
Merck

1.19258

Chlorine Powder Packs for photometers of other manufacturers for 25-ml samples

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


pH

6.3 (20 °C, 10 g/L in H2O)

Quality Segment

storage temp.

15-25°C





Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library