Skip to Content
Merck
All Photos(3)

Documents

07-1570-S

Sigma-Aldrich

Anti-monomethyl Histone H4 (Lys20) Antibody, Trial Size

Upstate®, from rabbit

Synonym(s):

H4K20me1, Histone H4 (monomethyl K20), H4 histone family, member A, Histone 1, H4a, Histone cluster 1, H4a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable
dot blot: suitable
inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

monomethylation (Lys20)

Gene Information

human ... H4C1(8359)

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. Histones modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. These modifications can alter local chromatin architecture, or recruit trans-acting factors that recognize specific histone modifications (the "histone code" hypothesis). Methylation of histone H4 at lysine 20 has been implicated in transcriptional activation, gene silencing, heterochromatin formation, mitosis, and DNA repair. Histone H4 is progressively methylated at Lys20 during the G(2), M, and G(1) phases of the cell cycle, and methylation of H4Lys20 is a marker for heterochromatin. In mammals, the female X chromosome is coated by the Xist RNA, which is critical for silencing, and is marked mainly by methylation of H3Lys9, H3Lys27 and H4Lys20.

Specificity

Based on sequence homology, broad species cross-reactivity is expected
Recognizes monomethylated Histone H4
(Lys20), Mr 11 kDa.

Immunogen

Epitope: Monomethyl Lys20
KLH-conjugated, linear synthetic peptide corresponding to amino acids 15-24
(AKRHR[me1K]VLRD-C) of human Histone H4.

Application

Anti-monomethyl Histone H4 (Lys20) Antibody, Trial Size is a rabbit polyclonal antibody for detection of monomethyl Histone H4 (Lys20) also known as H4K20me1, Histone H4 (mono methyl K20), H4 histone family member A & has been validated in WB.
Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (2 X 106 cell equivalents per IP) was subjected to chromatin immunoprecipitation using 4 μg of either a negative control antibody or Anti-Monomethyl-Histone H4
(Lys20) antibody and the Magna ChIP A Kit (Cat. #17-610). Successful immunoprecipitation of monomethyl-histone H4 (Lys20)-associated DNA fragments was verified by qPCR using GAPDH coding region ChIP Primers versus Control Primers corresponding to the GAPDH promoter.
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Dot Blot Analysis : Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665), which contain histone peptides with various modifications were probed with Cat. No. 07-1570-S Anti-monomethyl Histone H4 (Lys20) at 1:1000 dilution. Proteins were visualized using a Donkey anti-rabbit IgG conjugated to HRP and a chemiluminescence detection system.

Western Blot Analysis:
0.1 μg/mL of proteins were visualized using a goat anti-rabbit secondary antibody conjugated to HRP and a chemiluminescence detection system (Please see figures)

Peptide Blocking Assay:
40 μg of histone H4 peptide containing monomethyl Lys20 abolished detection of histone H4 by anti-monomethyl-Histone H4 (Lys20) in immunoblot analysis of acid extracts of HeLa cells (Please see figures).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Histones

Quality

Routinely evaluated by western blot analysis

Target description

~11 kDa

Physical form

Affinity Chromatography
Purified from serum by affinity chromatography. Supplied at 1 mg/ml in 0.1M Tris-Glycine (pH7.4), 15mM NaCl, 0.05% NaN3.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa cell acid extracts.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dmitry Karachentsev et al.
Developmental biology, 304(1), 46-52 (2007-01-19)
Methylation of specific amino acids in histone tails is responsible for packaging DNA into condensed, repressed chromatin, and into open chromatin that is accessible to the transcription machinery. Monomethylation and trimethylation of histone H4-lysine 20 (H4-K20) control the formation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service