bp
46/0.1mm
Iupac Name
3-phenoxy-2-butanone
Inchi Code
1S/C10H12O2/c1-8(11)9(2)12-10-6-4-3-5-7-10/h3-7,9H,1-2H3
InChI key
GYXROYABIGAUKY-UHFFFAOYSA-N
Purity
95%
Country of Origin
US
Product Link
https://www.matrixscientific.com/045804.html
Boiling point
46/0.1mm
MSDS
https://www.matrixscientific.com/045804.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.