Skip to Content
Merck
All Photos(1)

Key Documents

KEY235883526

Key Organics / BIONET

methyl 5-iodo-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
296.02
MDL number:

mp

283 - 285

Iupac Name

methyl 5-iodo-2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylate

Inchi Code

1S/C6H5IN2O4/c1-13-5(11)3-2(7)4(10)9-6(12)8-3/h1H3,(H2,8,9,10,12)

InChI key

GPLIKJXNEXUIGI-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/FD-0731

Melting Point

283 - 285

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library