ENAH95E72968
Enamine
4-(propa-1,2-dien-1-yl)piperidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-(propa-1,2-dien-1-yl)piperidine
Inchi Code
1S/C8H13N/c1-2-3-8-4-6-9-7-5-8/h3,8-9H,1,4-7H2
InChI key
DFBQZWCXLFMEEB-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-27144378
MSDS
https://www.enaminestore.com/catalog/EN300-27144378
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.