Skip to Content
Merck
All Photos(1)

Key Documents

ENAH039FE822

Enamine

2-(methoxymethyl)oxane-2-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
PLN 1,606.50
100 MG
PLN 2,302.65
250 MG
PLN 3,203.55
500 MG
PLN 4,917.60
1 G
PLN 6,251.40
2.5 G
PLN 12,054.60
5 G
PLN 17,734.95
10 G
PLN 26,205.75

Vendor SKU: EN300-1072672-50MG

PLN 1,606.50


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
PLN 1,606.50
100 MG
PLN 2,302.65
250 MG
PLN 3,203.55
500 MG
PLN 4,917.60
1 G
PLN 6,251.40
2.5 G
PLN 12,054.60
5 G
PLN 17,734.95
10 G
PLN 26,205.75

About This Item

CAS Number:
Molecular Weight:
174.2
MDL number:

Vendor SKU: EN300-1072672-50MG

PLN 1,606.50


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-(methoxymethyl)tetrahydro-2H-pyran-2-carboxylic acid

Inchi Code

1S/C8H14O4/c1-11-6-8(7(9)10)4-2-3-5-12-8/h2-6H2,1H3,(H,9,10)

InChI key

CWSWHKCJZSRKIB-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-1072672

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-1072672


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service