Skip to Content
Merck
All Photos(1)

Key Documents

ENA388700221

Enamine

5-chlorofuran-2-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
127.53
MDL number:

Iupac Name

5-chloro-2-furonitrile

Inchi Code

1S/C5H2ClNO/c6-5-2-1-4(3-7)8-5/h1-2H

InChI key

ZQXMLPGTNIYTRW-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-123911

Pictograms

CorrosionExclamation mark

Signal Word

Danger

MSDS

https://www.enaminestore.com/catalog/EN300-123911


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library