Skip to Content
Merck

COM448633864

Combi-Blocks, Inc.

4-Chloro-2-(N,N-dimethylaminomethyl)phenylboronic acid, pinacol ester

Synonym(s):

[5-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-N,N-dimethylmethanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C15H23O2N1Cl1B1
CAS Number:
Molecular Weight:
295.62
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H23BClNO2/c1-14(2)15(3,4)20-16(19-14)13-8-7-12(17)9-11(13)10-18(5)6/h7-9H,10H2,1-6H3

Inchi Key

YQQGIMWHFDQUNR-UHFFFAOYSA-N

Purity

98%

Storage temp.

Refrigerated

Country of Origin

CN



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library