Skip to Content
Merck
All Photos(1)

Key Documents

CIAH987EDA9A

ChemScene LLC (Preferred Partner)

2,6-Dichloropyridine-3,5-dicarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
198.01
MDL number:

bp

321.7+-37.0 C at 760 mmHg

Iupac Name

2,6-dichloro-3,5-pyridinedicarbonitrile

Inchi Code

1S/C7HCl2N3/c8-6-4(2-10)1-5(3-11)7(9)12-6/h1H

InChI key

JWVLIQKMDXWUFT-UHFFFAOYSA-N

storage temp.

4C

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/151229-84-4.html

Boiling point

321.7+-37.0 C at 760 mmHg

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0173578.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library