C12H316CF094
Chemcia Scientific, LLC
5-Fluoro-imidazo[1,2-a]pyridine hydrochloride
Select a Size
Vendor SKU: EN300-7542957-50MG
$408.20
Select a Size
About This Item
Vendor SKU: EN300-7542957-50MG
$408.20
Iupac Name
5-fluoroimidazo[1,2-a]pyridine hydrochloride
Inchi Code
1S/C7H5FN2.ClH/c8-6-2-1-3-7-9-4-5-10(6)7;/h1-5H;1H
InChI key
ZKQIOSYZRVPCKZ-UHFFFAOYSA-N
storage temp.
Salt Data
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
MSDS
http://www.chemcia.com/product_fl02.asp?id=6399
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.