Skip to Content
Merck

ATEH97A4DBEF

AstaTech, Inc.

1-(2,6-DICHLOROPHENYL)CYCLOBUTANAMINE HYDROCHLORIDE

Synonym(s):

1-(2,6-dichlorophenyl)cyclobutan-1-amine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H12N1Cl3
CAS Number:
Molecular Weight:
252.57
MDL number:
Product ATEH97A4DBEF is not currently sold in your country.


Inchi Code

1S/C10H11Cl2N.ClH/c11-7-3-1-4-8(12)9(7)10(13)5-2-6-10;/h1,3-4H,2,5-6,13H2;1H

Inchi Key

UQSSKVAWCTVEAX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

2-8 C

Country of Origin

US

Physical Form

Solid



signalword

Warning



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library