Skip to Content
Merck
All Photos(1)

Key Documents

ATEH95E592BD

AstaTech, Inc.

9-OXO-9,10-DIHYDROACRIDINE-4-CARBOXYLIC ACID

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
239.23
MDL number:

Iupac Name

9-oxo-9,10-dihydro-4-acridinecarboxylic acid

Inchi Code

1S/C14H9NO3/c16-13-8-4-1-2-7-11(8)15-12-9(13)5-3-6-10(12)14(17)18/h1-7H,(H,15,16)(H,17,18)

InChI key

BATAOFZEDHPRTM-UHFFFAOYSA-N

Purity

95%

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=156336

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=C15404


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library