ATEH3031F0B8
AstaTech, Inc.
3-BROMOBENZENEDIAZONIUM TETRAFLUOROBORATE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
3-bromobenzenediazonium tetrafluoroborate
Inchi Code
1S/C6H4BrN2.BF4/c7-5-2-1-3-6(4-5)9-8;2-1(3,4)5/h1-4H;/q+1;-1
InChI key
FIBPQIDZBMSYBA-UHFFFAOYSA-N
Purity
95%
Physical Form
Light-red to Brown Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=147046
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=98381
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.